missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Disposable IR Cards, 5x10cm, KBr window, 19mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

Manufacturer:  Alfa Aesar™ 43935KJ

 View more versions of this product

Catalog No. AA43935KJ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Size: 5 x 10cm
  • KBr window
  • 19mm aperture
Specifications

Specifications

Disposable IR Cards
5 Pk.
Documents