Honeywell™ Disposable Face Shields
Designed for rugged jobs—tough yet lightweight and comfortable
$26.90 - $259.96
Specifications
Certifications/Compliance | ANSI Z87.1-2010, CSA Z94.3 |
---|
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Product Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Coating | Lens Tint | Product Type | Price | Quantity | |||
29-635-3812
|
Honeywell™
UVS8500 |
Uncoated | Clear | Face Shield |
Each for $26.90
|
|
29-635-3813
|
Honeywell™
UVS8510 |
Anti-fog hardcoat | Clear | Face Shield |
Each for $44.16
|
|
29-635-3814
|
Honeywell™
UVS8550 |
Uncoated | Clear | Replacement visor |
Pack of 10 Each for $59.51
|
|
29-635-3817
|
Honeywell™
UVS8565 |
Uncoated | Shade 5 | Replacement visor |
Pack of 10 Each for $259.96
|
|
191300829
|
Honeywell™
UVS8555 |
Anti-fog hardcoat | Clear | Replacement visor |
Pack of 10 Each for $204.74
|
|
Description
For workers exposed to falling or flying objects, impact, chemical splash, and airborne debris. Fit with most goggles and respirators.
- Shell offers built-in chin protection and extended top-of-head coverage
- Adjustable headgear tilts visor into 2784 possible positions
- Ratcheting headgear features a locking mechanism for secure fit
- Comfortable cell foam on back of headgear with breathable, removable, washable headband
- 100% dielectric; no metal parts
Replacement Visors
- Clear lens suits most indoor work applications
- Shade 5.0 suits welding, brazing, and cutting
Compliance
Meets ANSI Z87.1+ and CSA Z94.3 standards.
Specifications
ANSI Z87.1-2010, CSA Z94.3 |