missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dynalon™ Dispensing Bottles

Low-density polyethylene bottles.

Manufacturer:  Dynalon™ 60508501EMD

 View more versions of this product

Catalog No. S31701




Description

Description

  • Supplied with a sealer cap that snaps on easily after using and seals tightly
  • Spout openings are standard 0.030 in. (0.08cm)
  • Opening can be enlarged by cutting back with scissors

Specifications

Specifications

1 oz., 30mL
1 oz.
3.82 in.
1.23 in.
No
Snap
30 mL
Red Cap
LDPE
31mm
97mm
Documents