missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium Rhodizonate 90.0+%, TCI America™

Manufacturer:  Tci America R001925G

 View more versions of this product

Catalog No. R001925G



Specifications

Specifications

Disodium Rhodizonate
300°C
523-21-7
MFCD00001650
SXWPCWBFJXDMAI-UHFFFAOYSA-L
disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
68225
Crystal-Powder at 20°C
Green-Yellow
25g
C6Na2O6
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
C1(=C(C(=O)C(=O)C(=O)C1=O)[O-])[O-].[Na+].[Na+]
214.04
214.04
≥90.0% (T)

Chemical Identifiers

523-21-7
214.04
SXWPCWBFJXDMAI-UHFFFAOYSA-L
68225
C1(=C(C(=O)C(=O)C(=O)C1=O)[O-])[O-].[Na+].[Na+]
C6Na2O6
MFCD00001650
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
Safety and Handling

Safety and Handling

TSCA : No

Documents