missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium Bathocuproine, For Copper, Certified, 0.1%, LabChem™

$284.86 - $284.86

Chemical Identifiers

CAS 7732-18-5,52698-84-7
CAS Max % 99.9,0.1
Linear Formula C26H18N2Na2O6S2
Formula Weight 564.54
Chemical Name or Material Disodium Bathocuproine, For Copper
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC136851 Lab Chem
LC136851
500mL Each for $284.86
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,52698-84-7
C26H18N2Na2O6S2
Disodium Bathocuproine, For Copper
99.9,0.1
564.54
Certified Grade

Specifications

564.54
Passes Test
Liquid
7732-18-5,52698-84-7
99.9,0.1
Room Temperature
Nitrogen oxides; Carbon monoxide; Carbon dioxide
0.1%
Soluble in water
Clear yellow
500mL
Passes Test
Poly Bottle
C26H18N2Na2O6S2
Disodium Bathocuproine, For Copper
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.