missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium 4-Formylbenzene-1,3-disulfonate, TCI America™

$62.64 - $456.81

Chemical Identifiers

CAS 33513-44-9
Molecular Formula C7H4Na2O7S2
Molecular Weight (g/mol) 310.202
MDL Number MFCD00043913
InChI Key UUKHCUPMVISNFW-UHFFFAOYSA-L
Synonym sodium 4-formylbenzene-1,3-disulfonate, benzaldehyde-2,4-disulfonic acid disodium salt, disodium 4-formylbenzene-1,3-disulfonate, disodium 4-formylbenzene-1,3-disulphonate, 1,3-benzenedisulfonic acid, 4-formyl-, disodium salt, 1,3-benzenedisulfonic acid, 4-formyl-, sodium salt 1:2, sodium 4-formylbenzene-1,3-disulfonate hydrate, 4-formyl-1,3-benzenedisulfonic acid, disodium salt, acmc-209i1i, benzaldehyde 2,4 disulfonic acid
PubChem CID 169512
IUPAC Name disodium;4-formylbenzene-1,3-disulfonate
SMILES C1=CC(=C(C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])C=O.[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B000825G View Documents TCI America
B000825G
25g Each for $62.64
 
B0008500G View Documents TCI America
B0008500G
500g Each for $456.81
 
Specifications

Chemical Identifiers

33513-44-9
310.202
UUKHCUPMVISNFW-UHFFFAOYSA-L
169512
C1=CC(=C(C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])C=O.[Na+].[Na+]
C7H4Na2O7S2
MFCD00043913
sodium 4-formylbenzene-1,3-disulfonate, benzaldehyde-2,4-disulfonic acid disodium salt, disodium 4-formylbenzene-1,3-disulfonate, disodium 4-formylbenzene-1,3-disulphonate, 1,3-benzenedisulfonic acid, 4-formyl-, disodium salt, 1,3-benzenedisulfonic acid, 4-formyl-, sodium salt 1:2, sodium 4-formylbenzene-1,3-disulfonate hydrate, 4-formyl-1,3-benzenedisulfonic acid, disodium salt, acmc-209i1i, benzaldehyde 2,4 disulfonic acid
disodium;4-formylbenzene-1,3-disulfonate

Specifications

White
33513-44-9
MFCD00043913
UUKHCUPMVISNFW-UHFFFAOYSA-L
disodium;4-formylbenzene-1,3-disulfonate
169512
Crystal-Powder at 20°C
Disodium 4-Formylbenzene-1,3-disulfonate
25g
C7H4Na2O7S2
sodium 4-formylbenzene-1,3-disulfonate, benzaldehyde-2,4-disulfonic acid disodium salt, disodium 4-formylbenzene-1,3-disulfonate, disodium 4-formylbenzene-1,3-disulphonate, 1,3-benzenedisulfonic acid, 4-formyl-, disodium salt, 1,3-benzenedisulfonic acid, 4-formyl-, sodium salt 1:2, sodium 4-formylbenzene-1,3-disulfonate hydrate, 4-formyl-1,3-benzenedisulfonic acid, disodium salt, acmc-209i1i, benzaldehyde 2,4 disulfonic acid
C1=CC(=C(C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])C=O.[Na+].[Na+]
310.202
310.20
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2108

TSCA : Yes