missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium 2,7-Naphthalenedisulfonate, TCI America™

$85.56 - $635.56

Chemical Identifiers

CAS 1655-35-2
Molecular Formula C10H6Na2O6S2
Molecular Weight (g/mol) 332.252
MDL Number MFCD00066365
InChI Key XOIWXJSPLXGSLZ-UHFFFAOYSA-L
Synonym 2,7-Naphthalenedisulfonic Acid Disodium Salt, Ebert-Merz alpha-Acid Disodium Salt
PubChem CID 74249
IUPAC Name disodium;naphthalene-2,7-disulfonate
SMILES C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0014500G View Documents TCI America
N0014500G
500g Each for $635.56
 
N001425G View Documents TCI America
N001425G
25g Each for $85.56
 
Specifications

Chemical Identifiers

1655-35-2
332.252
XOIWXJSPLXGSLZ-UHFFFAOYSA-L
74249
C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
C10H6Na2O6S2
MFCD00066365
2,7-Naphthalenedisulfonic Acid Disodium Salt, Ebert-Merz alpha-Acid Disodium Salt
disodium;naphthalene-2,7-disulfonate

Specifications

Undesignated
1655-35-2
MFCD00066365
XOIWXJSPLXGSLZ-UHFFFAOYSA-L
disodium;naphthalene-2,7-disulfonate
74249
Crystal-Powder at 20°C
Disodium 2,7-Naphthalenedisulfonate
500g
C10H6Na2O6S2
2,7-Naphthalenedisulfonic Acid Disodium Salt, Ebert-Merz alpha-Acid Disodium Salt
C1=CC(=CC2=C1C=CC(=C2)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
332.252
332.25
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0468

TSCA : Yes