missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Disodium 2,6-Naphthalenedisulfonate, TCI America™

$55.00 - $314.73

Chemical Identifiers

CAS 1655-45-4
Molecular Formula C10H6Na2O6S2
Molecular Weight (g/mol) 332.252
MDL Number MFCD00004092
InChI Key WZZLWPIYWZEJOX-UHFFFAOYSA-L
Synonym 2,6-naphthalenedisulfonic acid disodium salt, sodium naphthalene-2,6-disulfonate, disodium 2,6-naphthalenedisulfonate, disodium naphthalene-2,6-disulphonate, 2,6-naphthalenedisulfonic acid, disodium salt, ebert-merz beta-acid disodium salt, disodium naphthalene-2,6-disulfonate, dipotassium 2,6-naphthalenedisulfonate, acmc-209dsq, ksc910s4f
PubChem CID 3014027
IUPAC Name disodium;naphthalene-2,6-disulfonate
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N001325G View Documents TCI America
N001325G
25g Each for $55.00
 
N0013500G View Documents TCI America
N0013500G
500g Each for $314.73
 
Specifications

Chemical Identifiers

1655-45-4
332.252
WZZLWPIYWZEJOX-UHFFFAOYSA-L
3014027
C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1S(=O)(=O)[O-].[Na+].[Na+]
C10H6Na2O6S2
MFCD00004092
2,6-naphthalenedisulfonic acid disodium salt, sodium naphthalene-2,6-disulfonate, disodium 2,6-naphthalenedisulfonate, disodium naphthalene-2,6-disulphonate, 2,6-naphthalenedisulfonic acid, disodium salt, ebert-merz beta-acid disodium salt, disodium naphthalene-2,6-disulfonate, dipotassium 2,6-naphthalenedisulfonate, acmc-209dsq, ksc910s4f
disodium;naphthalene-2,6-disulfonate

Specifications

White
1655-45-4
MFCD00004092
WZZLWPIYWZEJOX-UHFFFAOYSA-L
disodium;naphthalene-2,6-disulfonate
3014027
Crystal-Powder at 20°C
Disodium 2,6-Naphthalenedisulfonate
25g
C10H6Na2O6S2
2,6-naphthalenedisulfonic acid disodium salt, sodium naphthalene-2,6-disulfonate, disodium 2,6-naphthalenedisulfonate, disodium naphthalene-2,6-disulphonate, 2,6-naphthalenedisulfonic acid, disodium salt, ebert-merz beta-acid disodium salt, disodium naphthalene-2,6-disulfonate, dipotassium 2,6-naphthalenedisulfonate, acmc-209dsq, ksc910s4f
C1=CC2=C(C=CC(=C2)S(=O)(=O)[O-])C=C1S(=O)(=O)[O-].[Na+].[Na+]
332.252
332.25
≥90.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0468

TSCA : No