Force of Nature Pro™ Disinfectant Storage Spray Bottles
Storage/spray bottles are reusable and Green Seal certified.
Manufacturer:  Force of Nature Pro™ FONSPRYBOTPRO001
Catalog No. 17000669
Description
Description
The Force of Nature Pro™ 32 oz. Spray Bottle has been designed both as a storage and spray bottle to maximize the 14 day product shelf life and maintain enough product inventory before another 32 oz. or 64 oz. batch can be made.
  • Force of Nature Pro™ product should be removed from the electrolyzer and stored in a spray bottle after each batch is made.
  • This item contains a one (1) Force of Nature Pro™ 32 oz. Spray Bottle
Specifications
Specifications
Dispenses disinfectant, sanitizer, deodorizer and multi-purpose cleaner
Storage/Spray Bottle
1
Force of Nature™ electrolyzer and capsules
Clear
32 oz.