Force of Nature™ Disinfectant Storage Spray Bottles
Storage/spray bottles are reusable and Green Seal certified.
Manufacturer:  Force of Nature™ FONSPRYBOT002
Catalog No. 17000666
Description
Description
Extra Force of Nature™ 12 oz. storage/spray bottles are ideal for any place you need to clean and deodorize safely.
  • Force of Nature™ bottles are heavy duty
  • Have fully adjustable spray nozzles (spray and stream)
  • Help maintain the Force of Nature™ solution shelf life after it is transformed by the electrolyzer appliance (sold separately)
    • The Force of Nature™ solution should be removed from the electrolyzer appliance and poured into 12 oz. Force of Nature™ bottles after the product has been transformed
Force of Nature™ spray bottles contain all the legal markings to comply with federal regulations.
Specifications
Specifications
Dispenses disinfectant, sanitizer, deodorizer and multi-purpose cleaner
Storage/Spray Bottle
1
Force of Nature™ electrolyzer and capsules
Clear
12 oz.