Please login to your online account to display your discounted pricing

Discovery Education™ Understanding Viruses DVD


Discusses viruses from the common cold to HIV, and how viral epidemics are more devastating than war.

Manufacturer: fisher science education  775585

 View more versions of this product

Catalog No. S68567

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Format VHS
Grade 6 to 12
Name Understanding: Viruses VHS
Run Time 54 min.
Copyright Year 1997