Please login to your online account to display your discounted pricing

Discovery Education™ Understanding: Bacteria VHS Video/DVD


Teach how bacteria both harm and help humans.

Manufacturer: fisher science education  775866A

 View more versions of this product

Catalog No. S68367DVD

  • / Each
 

Description & Specifications

Specifications

Grade 6 to 12
Name Understanding: Bacteria DVD
Run Time 54 min.
Copyright Year 2004
Includes Marc Record, Teacher's Guide
Format DVD