Please login to your online account to display your discounted pricing

Discovery Education™ Space Science: The Basics, Grades 3 to 5


From satellites and the atmosphere to GPS technology and the scientific method

Manufacturer: fisher science education  880575

 View more versions of this product

Catalog No. S90453DVD

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Copyright 2005
Name Space Science: the Basics 3-5 DVD
Run Time 180 min.
Grade 3 to 5
Format DVD
Includes Marc Record, Teacher's Guide