Please login to your online account to display your discounted pricing

Discovery Education™ Physical Science: The Basics, Grade K to 2


From electricity and magnetism to light and sound

Manufacturer: fisher science education  879213

 View more versions of this product

Catalog No. S90449

  • / Pack of 2 Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Format VHS
Grade K to 2
Name Physical Science: The Basics K-2 VHS
Run Time 180 min.
Copyright Year 2005