Please login to your online account to display your discounted pricing

Discovery Education™ MythBusters: Chemical Reactions DVD


Get out of getting “skunked.”

Manufacturer: fisher science education  902791

 View more versions of this product

Catalog No. S95459DVD

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Format DVD
Grade 6 to 12
Copyright Year 2007
Name Mythbusters: Chemical Reactions DVD
Run Time 16 min.