Please login to your online account to display your discounted pricing

Discovery Education™ Greatest Discoveries with Bill Nye: Chemistry


Host Bill Nye looks back over the past two centuries of Chemistry discoveries

Manufacturer: fisher science education  859033

 View more versions of this product

Catalog No. S87707DVD

  • / Each
 

Description & Specifications

Specifications

Format DVD
Copyright Year 2005
Includes Marc Record, Teacher's Guide
Grade 6 to 12
Name Greatest Discoveries with Bill Nye: Chemistry DVD
Run Time 45 min.