Please login to your online account to display your discounted pricing

Discovery Education™ Greatest Discoveries with Bill Nye: Chemistry


Host Bill Nye looks back over the past two centuries of Chemistry discoveries

Manufacturer: fisher science education  858951

 View more versions of this product

Catalog No. S87707

  • / Each
 

Description & Specifications

Specifications

Format VHS
Run Time 45 min.
Includes Marc Record, Teacher's Guide
Grade 6 to 12
Name Greatest Discoveries with Bill Nye: Chemistry VHS
Copyright Year 2005