missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Discovery Education™ Greatest Discoveries with Bill Nye: Chemistry

Host Bill Nye looks back over the past two centuries of Chemistry discoveries

$81.40 - $81.40

Specifications

Copyright Year 2005
Grade 6 to 12
Includes Marc Record, Teacher's Guide
Run Time 45 min.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Name Format Price Quantity  
Catalog Number Mfr. No. Name Format Price Quantity  
S87707DVD Fisher Science Education
859033
Greatest Discoveries with Bill Nye: Chemistry DVD DVD Each for $81.40
Specifications

Description

Nye examines the second half of the 19th century, a time dominated by discoveries relating to light, electrons, radioactivity, and the periodic table of the elements as well as 20th century advances in science. Running time: 45 min.

Topics include:

  • How electricity transforms chemicals
  • How elements can combine into more complex molecules
  • How the combination of nonliving substances produced organic compounds that led to pharmacology
  • Nye examines the second half of the 19th century, a time dominated by discoveries relating to light, electrons, radioactivity, and the periodic table of the elements as well as 20th century advances in science

Description & Specifications

Specifications

2005
6 to 12
Marc Record, Teacher's Guide
45 min.