Please login to your online account to display your discounted pricing

Discovery Education™ Greatest Discoveries with Bill Nye: Biology


Join Bill Nye as he delves into the Greatest Discoveries in Biology

Manufacturer: fisher science education  859058

 View more versions of this product

Catalog No. S87706DVD

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Format DVD
Grade 6 to 12
Name Greatest Discoveries with Bill Nye: Biology DVD
Run Time 45 min.
Copyright Year 2005