Please login to your online account to display your discounted pricing

Discovery Education™ Great Inventions with Bill Nye: Science of Materials


See the history and technology of the materials around us

Manufacturer: fisher science education  903716

 View more versions of this product

Catalog No. S95449DVD

  • / Each
 

Description & Specifications

Specifications

Copyright Year 2007
Name Greatest Inventions with Bill Nye: The Science Of Materials DVD
Format DVD
Grade 6 to 12
Run Time 56 min.
Includes Marc Record, Teacher's Guide