missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Discovery Education™ Forensics CD-ROM

Learn what occurs during crime scene investigations — how data is sorted, how tests are run to determine the chemical makeup of evidence, and how the importance of items is decided.

$145.33 - $145.33

Specifications

Compatibility Windows™ 2000, XP™ and Mac™ OSX
Category Chemistry
Format CD-ROM
Product Type Forensics
For Use With (Application) Determine the chemical makeup of evidence and decide on the importance of items
View More Specs

Products 1
Catalog Number Mfr. No. License Price Quantity  
Catalog Number Mfr. No. License Price Quantity  
S68416A View Documents Discovery Education™
774166A
Single User Each for $145.33
 
Description

Description

  • Science Simulations: Students can perform hundreds of simulated lab experiments like Crime Scene Investigation, Firefighting and Scientific Inquiry
  • Discovery Channel™ Theater: Five fast-paced, fact-filled video voyages (Crime Scenes, DNA, Forgery, Mummies and Arson) in both English and Spanish
  • Library: World Book™ Encyclopedia with extensive reference and research materials
  • Laboratory: Full of activities to build critical learning skills
  • Multimedia Studio: Tool with slides, text and sound effects allows students to create their own presentations (also includes Lesson Launchers — ready-made slide shows to begin teaching the next science topic)
  • Teacher's guide on CD-ROM has more than 100 pages of support material; also includes free online lesson support
  • Updated, user-friendly, and age-appropriate
  • Single-user CD-ROM (Version 2.0); Windows™ 2000, Windows XP™ and Macintosh™ OS X compatible
  • Grades 5–9

Specifications

Specifications

Windows™ 2000, XP™ and Mac™ OSX
CD-ROM
Determine the chemical makeup of evidence and decide on the importance of items
Teacher's guide of more than 100 pages of support material also includes free online lesson support
Chemistry
Forensics
12