Please login to your online account to display your discounted pricing

Discovery Education™ Energy Alternatives Cool Fuel Road Trips DVDs

Follow Shaun Murphy and the Cool Fuel crew as they take alternate energy solutions on the road.

Manufacturer: fisher science education  904219

 View all options for this product

Catalog No. S95985DVD


Description & Specifications


Includes Marc Record, Teacher's Guide
Format DVD
Grade 6 to 12
Name Energy Alternatives: The Cool Fuel Road Trips: Water and Wind Power DVD
Run Time 23 min.

Geothermal and Solar Energy

  • Shaun and the crew leave their gas tanks in San Francisco during a geothermal-energy-powered trek to the Oregon border
  • Later, Shaun and his dog Sparky rely on solar power while chasing the sun through Arizona
  • Run time: 25 min.
  • Grades 6 to 12
Water and Wind Power
  • Shaun and the crew make a 250-mile trip through Colorado fueled by micro-hydropower to demonstrate that energy can be generated from small streams
  • Later, Shaun tries to catch the wind with turbine power for a journey along Route 66 in New Mexico
  • Run time: 23 min.
  • Grades 6 to 12