Please login to your online account to display your discounted pricing

Discovery Education™ Elements of Physics: Light DVD


Learn about the difficulties scientists have had in unraveling the secrets of this fundamental force.

Manufacturer: fisher science education  895722

 View more versions of this product

Catalog No. S95995DVD

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Format DVD
Grade 6 to 12
Name Elements Of Physics: Light DVD
Run Time 56 min.