Please login to your online account to display your discounted pricing

Discovery Education™ Elements of Earth Science Series VHS/DVD


Four videos explore the basic elements of earth science including geology, meteorology, and hydrology.

Manufacturer: fisher science education  880799

 View more versions of this product

Catalog No. S95992DVD

  • / Each
 

Description & Specifications

Specifications

Grade 3 to 5
Name Rocks, Minerals and Soils DVD
Copyright Year 2005
Format DVD
Run Time 30 min.
Includes Marc Record, Teacher's Guide