Please login to your online account to display your discounted pricing

Discovery Education™ Elements of Chemistry DVD Set


Six DVDs explore the essentials of chemistry.

Manufacturer: fisher science education  895631

 View more versions of this product

Catalog No. S94005DVD

  • / Each
 

Description & Specifications

Specifications

Format DVD
Run Time 336 min.
Grade 6 to 12
Name Elements of Chemistry 6-pack DVD
Includes Marc Record