Please login to your online account to display your discounted pricing

Discovery Education™ Elements of Biology


Explore the world of living organisms.

Manufacturer: fisher science education  DVD895631

 View more versions of this product

Catalog No. S94004DVD

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record
Format DVD
Grade 6 to 12
Name Elements of Biology 6-pack DVD
Run Time 336 min.