Please login to your online account to display your discounted pricing

Discovery Education™ Elements of Biology


Explore the world of living organisms.

Manufacturer: fisher science education  VHS894873

 View more versions of this product

Catalog No. S94004

  • / Each
 

Description & Specifications

Specifications

Name Elements of Biology 6-pack VHS
Includes Marc Record
Format VHS
Run Time 336 min.
Grade 6 to 12