missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Discovery Education™ Earth Science: The Basics, Grade K to 2

From oceans and volcanoes to fossils and storms

Manufacturer:  Discovery Education™ 880500

 View more versions of this product

Catalog No. S90446DVD



Description

Description

Introduce young students to a wide variety of Earth science topics with these segments drawn from this award-winning series.

The title includes six scientific literacy segments to aid classroom teachers along with the following 12 student video segments:

  • Protecting Earth
  • Earth's Atmosphere
  • Weather and Seasons
  • Storms
  • Dinosaurs and Fossils
  • Rocks: An Introduction
  • Land & Water: An Introduction
  • Cave Formation
  • Rivers
  • Oceans: Introduction
  • Volcanoes: Introduction
  • Investigating Earthquakes

Order Info

Grade K to 2. Running time: 3 hours.

Specifications

Specifications

2005
Earth Science: The Basics Grades K-2 DVD
DVD
K
Marc Record, Teacher's Guide
180 min.
2
Education
Documents