Please login to your online account to display your discounted pricing

Dirhenium decacarbonyl, sublimed, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dirhenium decacarbonyl
CAS 14285-68-8
Molecular Formula C10O10Re2
Linear Formula Re2(CO)10
Formula Weight 652.51
MDL Number MFCD00011198
Synonym Rhenium carbonyl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC223990050

  SDS
thermo fisher scientific
223990050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 652.51
Color Light greenish-yellow
Quantity 5g
EINECS Number 238-202-8
TSCA TSCA
Chemical Name or Material Dirhenium decacarbonyl
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C10O10Re2
Linear Formula Re2(CO)10
MDL Number MFCD00011198
CAS 14285-68-8
Synonym Rhenium carbonyl
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
Call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.