missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440590250

 View more versions of this product

Catalog No. AC440590250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Coupling reagent
Specifications

Specifications

Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate
Authentic
White
25g
97%
1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 2,5-dioxopyrrolidin-1-yl oxy bis pyrrolidin-1-yl methylium hexafluorophosphate, bis pyrrolidin-1-yl methylidene 2,5-dioxopyrrolidin-1-yl oxidanium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate chn, hspyu, n,n,n',n'-bis tetramethylene-o-n-succinimidyl uronium hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)ON3C(=O)CCC3=O.F[P-](F)(F)(F)(F)F
411.29
411.29
97%
36.45 to 39.48% C, 9.81 to 10.63% N
Glass bottle
141°C to 143.5°C
207683-26-9
C13H20N3O3·F6P
TVWKVGNCIJUXMY-UHFFFAOYSA-N
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]pyrrolidine-2,5-dione;hexafluorophosphate
16218611
Powder or Crystalline Powder

Chemical Identifiers

207683-26-9
411.29
1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 2,5-dioxopyrrolidin-1-yl oxy bis pyrrolidin-1-yl methylium hexafluorophosphate, bis pyrrolidin-1-yl methylidene 2,5-dioxopyrrolidin-1-yl oxidanium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate chn, hspyu, n,n,n',n'-bis tetramethylene-o-n-succinimidyl uronium hexafluorophosphate
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]pyrrolidine-2,5-dione;hexafluorophosphate
C13H20N3O3·F6P
TVWKVGNCIJUXMY-UHFFFAOYSA-N
16218611
C1CCN(C1)C(=[N+]2CCCC2)ON3C(=O)CCC3=O.F[P-](F)(F)(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents