Please login to your online account to display your discounted pricing

Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate
Assay Percent Range 97%
Molecular Formula C13H20N3O3·F6P
Formula Weight 411.29
Synonym HSPyU
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC440590250

  SDS
thermo fisher scientific
AC440590250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440590050

  SDS
thermo fisher scientific
AC440590050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Formula Weight 411.29
Quantity 25g
Color White to off-white
Chemical Name or Material Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate
Packaging Glass bottle
Molecular Formula C13H20N3O3·F6P
CAS 207683-26-9
Synonym HSPyU
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Elemental Analysis 36.45 to 39.48% C, 9.81 to 10.63% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Coupling reagent

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.