Please login to your online account to display your discounted pricing

Dipropyl phthalate, +99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  409160050

 View more versions of this product

Catalog No. AC409160050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Density 1.0780g/mL
Color Clear colorless
Quantity 5g
Chemical Name or Material Dipropyl phthalate
CAS 131-16-8
Physical Form Clear colorless liquid
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C14H18O4
Formula Weight 250.29g/mol
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Dipropyl phthalate
Assay 99.0% min.
Assay Percent Range 99+%
Molecular Formula C14H18O4
Formula Weight 250.29g/mol
Accessories

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.

SDS & Safety Documents