Please login to your online account to display your discounted pricing

DIPPA hydrochloride, Tocris


Selective irreversible κ antagonist

Quantity 10mg

Manufacturer: r & d systems  079410

 View more versions of this product

Catalog No. 079410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 484.87
Chemical Name or Material DIPPA hydrochloride
Assay Percent Range >99%
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(3-isothiocyanatophenyl)-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Recommended Storage Desiccate at -20°C
Molecular Formula C22H23Cl2N3OS·HCl
CAS 155512-52-0

Specifications

Chemical Name or Material DIPPA hydrochloride
Assay Percent Range >99%
Molecular Formula C22H23Cl2N3OS·HCl
Formula Weight 484.87
Synonym 2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(3-isothiocyanatophenyl)-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
Accessories

Desiccate at -20°C