Please login to your online account to display your discounted pricing

Alfa Aesar™ Dipinacolyl methylphosphonate, 97%

$72.93 - $849.81

Chemical Identifiers

Chemical Name or Material Dipinacolyl methylphosphonate
CAS 7040-58-6
Molecular Formula C13H29O3P
Formula Weight 264.35
MDL Number MFCD00440711
Synonym Methylphosphonic acid bis(3,3-dimethyl-2-butyl) ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4464002

  SDS
alfa aesar™
4464002
0.5g Each for $72.93

AA4464009

  SDS
alfa aesar™
4464009
10g Each for $849.81
Description & Specifications

Specifications

Formula Weight 264.35
Boiling Point 75°C (0.8mmHg)
Storage Note 1
  • Packaged under Argon
Physical Form Liquid
Molecular Formula C13H29O3P
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 0.5g
Chemical Name or Material Dipinacolyl methylphosphonate
Assay 97%
Sensitivity Air & Moisture Sensitive
CAS 7040-58-6
Health Hazard 1 Warning
Synonym Methylphosphonic acid bis(3,3-dimethyl-2-butyl) ester
MDL Number MFCD00440711

  • Packaged under Argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Packaged under Argon