missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dipinacolyl methylphosphonate, 97%

Manufacturer:  Alfa Aesar™ 4464004

 View more versions of this product

Catalog No. AA4464004



Description

Description

  • Packaged under Argon
Specifications

Specifications

Dipinacolyl methylphosphonate
75°C (0.8mmHg)
7040-58-6
MFCD00440711
XSQAITMTGIQAHD-UHFFFAOYSA-N
3-[3,3-dimethylbutan-2-yloxy(methyl)phosphoryl]oxy-2,2-dimethylbutane
23454
Liquid
Air and moisture sensitive
2g
C13H29O3P
3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimet
CC(C(C)(C)C)OP(=O)(C)OC(C)C(C)(C)C
264.346
264.35
97%

Chemical Identifiers

7040-58-6
264.346
XSQAITMTGIQAHD-UHFFFAOYSA-N
23454
CC(C(C)(C)C)OP(=O)(C)OC(C)C(C)(C)C
C13H29O3P
MFCD00440711
3-3,3-dimethylbutan-2-yloxy methyl phosphoryl oxy-2,2-dimet
3-[3,3-dimethylbutan-2-yloxy(methyl)phosphoryl]oxy-2,2-dimethylbutane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Documents