missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenylzinc, 98+%

Manufacturer:  Alfa Aesar™ 8790803

 View more versions of this product

Catalog No. AA8790803



Specifications

Specifications

Diphenylzinc
1078-58-6
280°C to 285°C
C12H10Zn
UN3181
BUNROVZNGITPIX-UHFFFAOYSA-N
zinc;benzene
517896
Crystalline
107°C
1g
Air & Moisture Sensitive
MFCD00014071
diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2, diphenylzinc
C1=CC=[C-]C=C1.C1=CC=[C-]C=C1.[Zn+2]
219.592
219.58
≥98%

Chemical Identifiers

1078-58-6
219.592
BUNROVZNGITPIX-UHFFFAOYSA-N
517896
C1=CC=[C-]C=C1.C1=CC=[C-]C=C1.[Zn+2]
C12H10Zn
MFCD00014071
diphenylzinc, zinc, diphenyl, mkrvhlwavkjbfn-uhfffaoysa-n, diphenyl zinc, diphenylzink, znph2, diphenylzinc
zinc;benzene
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

SDS Documents