missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyltin dichloride, 95%

Manufacturer:  Alfa Aesar™ 7113918

 View more versions of this product

Catalog No. AA7113918



Specifications

Specifications

Diphenyltin dichloride
41°C to 43°C
50g
White
C12H10Cl2Sn
MFCD00000516
ISXUHJXWYNONDI-UHFFFAOYSA-L
dichloro(diphenyl)stannane
14342
Crystalline
333°C to 337°C (decomposition)
1135-99-5
>150°C (302°F)
Moisture Sensitive
(C6H5)2SnCl2
diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(Cl)Cl
343.822
343.81
95%

Chemical Identifiers

1135-99-5
343.822
ISXUHJXWYNONDI-UHFFFAOYSA-L
14342
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(Cl)Cl
C12H10Cl2Sn
MFCD00000516
diphenyltin dichloride, stannane, dichlorodiphenyl, dichlorodiphenyltin, diphenyldichlorotin, diphenyltin chloride, diphenylstannyl dichloride, tin, diphenyl-, dichloride, dichloro diphenyl stannane, diphenylstannium dichloride, unii-0y7mz4k0dr
dichloro(diphenyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : WH7253000

SDS Documents