Please login to your online account to display your discounted pricing

Diphenylthiocarbazone Solution, LabChem™

$158.29 - $158.29

Chemical Identifiers

Chemical Name or Material Diphenylthiocarbazone Solution
CAS 67-66-3
CAS Max % 98.9901
CAS 64-17-5
CAS Max % 0.9999
CAS 60-10-6
CAS Max % 0.01
Linear Formula C13H12N4S
Formula Weight 256.3
UN Number UN1888
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC136882

 
lab chem
LC136882
1L Each for $158.29
Description & Specifications

Specifications

Formula Weight 256.3
Chemical Name or Material Diphenylthiocarbazone Solution
CAS Max % 0.01
Decomposition Information Chlorine; Carbon monoxide; Carbon dioxide; Nitrogen oxides
Boiling Point 61°C
Color Green-black
Melting Point −64°C
Quantity 1L
Physical Form Liquid
Linear Formula C13H12N4S
Density 1,485kg/m3
CAS 67-66-3
UN Number UN1888
Synonym Dithizone
Health Hazard 1 Warning
CAS Max % 98.9901
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
Toxic to aquatic life.
Identification Passes Test
CAS 64-17-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Amber Glass
CAS Max % 0.9999
Solubility Soluble in water
CAS 60-10-6
Specific Gravity 1485kg/m3
Storage Information Room Temperature
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
Toxic to aquatic life.