Please login to your online account to display your discounted pricing

Diphenylthiocarbazone, ACS Grade, LabChem™

$153.57 - $153.57

Chemical Identifiers

Chemical Name or Material Diphenylthiocarbazone
CAS 60-10-6
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C13H12N4S
Formula Weight 256.3
Synonym Dithizone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC136877

 
lab chem
LC136877
Amber Glass 10g Each for $153.57
Description & Specifications

Specifications

Formula Weight 256.3
Chemical Name or Material Diphenylthiocarbazone
EINECS Number 200-454-1
Packaging Amber Glass
Solubility Soluble in water
Physical Form Powder
Suitability Passes Test
CAS 60-10-6
Synonym Dithizone
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Identification Passes Test
Recommended Storage Room Temperature
Color Black
Melting Point 168°C
Quantity 10g
Purity Grade ACS Grade
Linear Formula C13H12N4S
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Room Temperature