missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylphosphoryl azide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC409041000

 View more versions of this product

Catalog No. AC409041000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

275.19
≥97.5%
Clear yellow to amber
04,210; 05,280; 07,138; 08,211; 10,173
C12H10N3O3P
MFCD00001987
Glass bottle
98%
Liquid
100g
1.2730g/mL
1.5500 to 1.5520 (20°C, 589nm)
26386-88-9
DPPA
Diphenylphosphoryl azide
Authentic

Chemical Identifiers

26386-88-9
C12H10N3O3P
MFCD00001987
Diphenylphosphoryl azide
98%
275.19
DPPA
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.
Toxic if inhaled.
May cause respiratory irritation.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger