Please login to your online account to display your discounted pricing

Alfa Aesar™ (Diphenylphosphonimido)triphenylphosphorane, 98%

$96.28 - $354.09

Chemical Identifiers

Chemical Name or Material (Diphenylphosphonimido)triphenylphosphorane
CAS 2156-69-6
Molecular Formula C30H25NOP2
Formula Weight 477.49
MDL Number MFCD00045685
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1758206

  SDS
alfa aesar™
L1758206
5g Each for $96.28

AAL1758214

  SDS
alfa aesar™
L1758214
25g Each for $354.09
Description & Specifications

Specifications

Formula Weight 477.49
Assay 98%
Molecular Formula C30H25NOP2
MDL Number MFCD00045685
Quantity 5g
Melting Point 170° to 174°C
CAS 2156-69-6
Chemical Name or Material (Diphenylphosphonimido)triphenylphosphorane

Applications
(Diphenylphosphonimido)triphenylphosphorane is used as a ligand and form coordination complexes with metal ions in synthetic chemistry.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.