missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylphosphonic azide, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1212414

 View more versions of this product

Catalog No. AAA1212414



Description

Description

Applications
Diphenylphosphonic azide acts as a reagent for the synthesis of peptides and phosphoramidates by reacting with amines. It is also used in the preparation of oligosaccharides linked with carbamate and urea bonds utilizing modified Curtis rearrangement. It is involved in pseudohalogen replacement of the azido group by treatment with nucleophilic reagents, such as water, butanol, ammonia, and various amines. Further, it is used as a hydroazidation catalyst for preparation of organoazides.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids and oxidizing agents.
Specifications

Specifications

Diphenylphosphonic Azide
25g
97%
MFCD00001987
2058967
SORGEQQSQGNZFI-UHFFFAOYSA-N
[azido(phenoxy)phosphoryl]oxybenzene
123414
Liquid
1.275
26386-88-9
C12H10N3O3P
UN3278
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av
C1=CC=C(C=C1)OP(=O)(N=[N+]=[N-])OC2=CC=CC=C2
275.204
275.2
97%

Chemical Identifiers

26386-88-9
275.204
SORGEQQSQGNZFI-UHFFFAOYSA-N
123414
C1=CC=C(C=C1)OP(=O)(N=[N+]=[N-])OC2=CC=CC=C2
C12H10N3O3P
MFCD00001987
diphenylphosphoryl azide, diphenyl azidophosphate, diphenylphosphonic azide, diphenyl phosphoryl azide, diphenyl phosphorazidate, phosphorazidic acid, diphenyl ester, azido phenoxy phosphoryl oxy benzene, dppa polymer-bound, diphenylphosphorazidate, unii-gxm91165av
[azido(phenoxy)phosphoryl]oxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P361-P304+P340-P311-P362-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.

Recommended Storage : 2°C to 8°C

SDS Documents