Please login to your online account to display your discounted pricing

Diphenylphosphinyl chloride, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  212960100

 View more versions of this product

Catalog No. AC212960100

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 236.64
Quantity 10g
Color Clear colorless to yellow
EINECS Number 216-107-2
Density 1.24g/mL
Beilstein 16, II, 386
Fieser 07,138
Chemical Name or Material Diphenylphosphinyl chloride
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Refractive Index 1.6080 to 1.6120 (20°C, 589nm)
Molecular Formula C12H10ClOP
Linear Formula (C6H5)2P(O)Cl
CAS 1499-21-4
Synonym Chlorodiphenylphosphine oxide
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 98%
MDL Number MFCD00002077

Specifications

Chemical Name or Material Diphenylphosphinyl chloride
Name Note 98%
CAS 1499-21-4
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H10ClOP
Linear Formula (C6H5)2P(O)Cl
Formula Weight 236.64
MDL Number MFCD00002077
Synonym Chlorodiphenylphosphine oxide
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

SDS & Safety Documents