missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylphosphinic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1156306

 View more versions of this product

Catalog No. AAA1156306



Description

Description

Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylphosphinic acid is used as a reagent employed in the synthesis of bidentate ligands and peptide coupling agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Diphenylphosphinic acid
193°C to 197°C
MFCD00002132
2804567
Soluble in water.
OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
218.19
CHEBI:37832
99%
1707-03-5
C12H11O2P
5 g
phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
BEQVQKJCLJBTKZ-UHFFFAOYSA-N
diphenylphosphinic acid
15567
218.19

Chemical Identifiers

1707-03-5
218.19
BEQVQKJCLJBTKZ-UHFFFAOYSA-N
15567
diphenylphosphinic acid
C12H11O2P
MFCD00002132
phosphinic acid, diphenyl, hydroxydiphenylphosphine oxide, diphenylphosphinicacid, diphenyl phosphinic acid, diphenyl-phosphinic acid, acmc-1boli, di phenyl phosphinic acid, p,p-diphenylphosphinic acid, ksc180o2h, phosphinicacid, p,p-diphenyl
CHEBI:37832
OP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-948-5

RTECSNumber : SZ5315000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only