missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylphosphine, Alfa Aesar™

Manufacturer:  Alfa Aesar 5616918

 View more versions of this product

Catalog No. AA5616918



Specifications

Specifications

Diphenylphosphine
110°C (230°F)
Light, Air & Moisture Sensitive
Colorless
829-85-6
MFCD00003040
diphenylphosphine, phosphine, diphenyl, ph2ph, unii-f9b5t7o7zy, diphenyl phosphine, hpph2, f9b5t7o7zy, diphenylphosphine 10wt%inhexane, diphenylphospine, diphenyl-phosphine
C1=CC=C(C=C1)PC2=CC=CC=C2
186.194
186.2
1.07g/mL
Unpleasant
280°C
50g
C12H11P
UN2845
GPAYUJZHTULNBE-UHFFFAOYSA-N
diphenylphosphane
70017
Liquid

Chemical Identifiers

829-85-6
186.194
GPAYUJZHTULNBE-UHFFFAOYSA-N
70017
C1=CC=C(C=C1)PC2=CC=CC=C2
C12H11P
MFCD00003040
diphenylphosphine, phosphine, diphenyl, ph2ph, unii-f9b5t7o7zy, diphenyl phosphine, hpph2, f9b5t7o7zy, diphenylphosphine 10wt%inhexane, diphenylphospine, diphenyl-phosphine
diphenylphosphane
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H315-H319-H335 Catches fire spontaneously if exposed to air. Self-heating in large quantities may catch fire. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
P210-P222-P305+P351+P338-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

SDS Documents