missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylmethylphosphine, polymer-supported, 0.9-1.4 mmol/g on polystyrene, Thermo Scientific Chemicals
$213.05 - $213.05
Chemical Identifiers
CAS | 1486-28-8 |
---|---|
Molecular Formula | C13H13P |
Molecular Weight (g/mol) | 200.22 |
MDL Number | MFCD00008508 |
InChI Key | UJNZOIKQAUQOCN-UHFFFAOYSA-N |
Synonym | methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine |
PubChem CID | 73879 |
IUPAC Name | methyl(diphenyl)phosphane |
SMILES | CP(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1947706
|
Thermo Scientific Chemicals
L1947706 |
5 g |
Each for $213.05
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1486-28-8 | |
200.22 | |
UJNZOIKQAUQOCN-UHFFFAOYSA-N | |
73879 | |
CP(C1=CC=CC=C1)C1=CC=CC=C1 |
C13H13P | |
MFCD00008508 | |
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine | |
methyl(diphenyl)phosphane |
Specifications
1486-28-8 | |
MFCD00008508 | |
5 g | |
CP(C1=CC=CC=C1)C1=CC=CC=C1 | |
200.22 | |
Diphenylmethylphosphine, polymer-supported, 0.9 to 1.4mmol/g on polystyrene |
C13H13P | |
methyldiphenylphosphine, diphenylmethylphosphine, methyl diphenyl phosphine, phosphine, methyldiphenyl, methyldiphenylphsphine, diphenylphosphinomethane, methyldiphenylphoshine, pubchem6421, methyl diphenylphosphine, methyldi-phenylphosphine | |
UJNZOIKQAUQOCN-UHFFFAOYSA-N | |
methyl(diphenyl)phosphane | |
73879 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only