missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylmethane, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 117350010
Specifications
Diphenylmethane | |
127°C | |
Glass bottle | |
1 | |
Colorless to Yellow | |
1kg | |
101-81-5 | |
C13H12 | |
MFCD00004781 | |
15, 3362 | |
Solubility in water: practically insoluble. Other solubilities: insoluble in liquid ammonia, freely soluble in alcohol, ether, chloroform,, hexane and benzene | |
C1=CC=C(C=C1)CC2=CC=CC=C2 | |
168.24 | |
CHEBI:38884 | |
Low Melting Solid |
1.0000g/mL | |
Authentic | |
1.5760 to 1.5780 | |
265.0°C | |
24.5°C to 25.0°C | |
99% | |
98.5% min. (GC) | |
(C6H5)2CH2 | |
05, II, 498 | |
1,1'-dimethylenebis benzene, benzene, 1,1'-methylenebis, benzene, benzyl, benzene, phenylmethyl, diphenyl methane, diphenylmethane, ditan, ditane, methane, diphenyl, methylenedibenzene | |
CZZYITDELCSZES-UHFFFAOYSA-N | |
benzylbenzene | |
7580 | |
168.24 | |
99% |
Chemical Identifiers
101-81-5 | |
168.24 | |
CZZYITDELCSZES-UHFFFAOYSA-N | |
7580 | |
benzylbenzene |
C13H12 | |
MFCD00004781 | |
1,1'-dimethylenebis benzene, benzene, 1,1'-methylenebis, benzene, benzyl, benzene, phenylmethyl, diphenyl methane, diphenylmethane, ditan, ditane, methane, diphenyl, methylenedibenzene | |
CHEBI:38884 | |
C1=CC=C(C=C1)CC2=CC=CC=C2 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 202-978-6
RTECSNumber : DA4976500
TSCA : TSCA