missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylmethane, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2125622

 View more versions of this product

Catalog No. AAB2125622



Description

Description

Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylmethane, is used as an adhesive chemical composition for making flexible laminates for use in food packaging.

Solubility
Soluble in ethanol, ether, benzene and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Incompatible with strong oxidizing agents.
Specifications

Specifications

Diphenylmethane
22°C to 26°C
262°C to 264°C
C13H12
MFCD00004781
UN3077
14,3328
Soluble in ethanol,ether,benzene and chloroform.
C(C1=CC=CC=C1)C1=CC=CC=C1
168.24
CHEBI:38884
≥99%
101-81-5
1.01
130°C (266°F)
1.577
100 g
1904982
diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
CZZYITDELCSZES-UHFFFAOYSA-N
benzylbenzene
7580
168.24

Chemical Identifiers

101-81-5
168.24
CZZYITDELCSZES-UHFFFAOYSA-N
7580
benzylbenzene
C13H12
MFCD00004781
diphenylmethane, ditan, ditane, benzene, 1,1'-methylenebis, diphenyl methane, benzene, benzyl, methane, diphenyl, 1,1'-dimethylenebis benzene, benzene, phenylmethyl, methylenedibenzene
CHEBI:38884
C(C1=CC=CC=C1)C1=CC=CC=C1
Safety and Handling

Safety and Handling

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 202-978-6

RTECSNumber : DA4976500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only