Please login to your online account to display your discounted pricing

Alfa Aesar™ Diphenyliodonium trifluoromethanesulfonate, 99%

Chemical Identifiers

Chemical Name or Material Diphenyliodonium trifluoromethanesulfonate
CAS 66003-76-7
Molecular Formula C13H10F3IO3S
Formula Weight 430.19
MDL Number MFCD00191356
UN Number UN2811
Beilstein 3582403
Synonym Diphenyliodonium triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1744403

 
alfa aesar™
L1744403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 430.19
Chemical Name or Material Diphenyliodonium trifluoromethanesulfonate
Assay 99%
Molecular Formula C13H10F3IO3S
CAS 66003-76-7
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00191356
UN Number UN2811
Beilstein 3582403
Synonym Diphenyliodonium triflate
Health Hazard 1 Danger
Quantity 1g
Melting Point 179° to 182°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.