Please login to your online account to display your discounted pricing

Diphenyliodonium nitrate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diphenyliodonium nitrate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H10I·NO3
Linear Formula (C6H5)2I+(NO3)-
Formula Weight 343.11
MDL Number MFCD00031704
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC292340050

  SDS
thermo fisher scientific
292340050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 343.11
Melting Point >147°C
Color White to grayish
Physical Form Fine Crystalline Powder
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C12H10I·NO3
Chemical Name or Material Diphenyliodonium nitrate
Assay Percent Range 97%
MDL Number MFCD00031704
CAS 722-56-5
Linear Formula (C6H5)2I+(NO3)-